Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MP Biomedicals Bovine Adult Serum
Used for the cultivation of Helicobacter pylori and in various low-serum media for diploid cell culture
Specifications
Color | Golden Brown |
---|---|
Format | Liquid |
Product Type | Bovine Serum |
Sterility | Sterile |
For Use With (Application) | Cultivation of Helicobacter pylori and in various low Serum media for diploid cell culture |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
ICN19135291
|
MP Biomedicals
0219135291 |
1L |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
ICN19135280
|
MP Biomedicals
0219135280 |
100mL |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
Description
Prices may fluctuate. Contact your Fisher Scientific Customer Service Representative for details.
Specifications
Golden Brown | |
Bovine Serum | |
Cultivation of Helicobacter pylori and in various low Serum media for diploid cell culture | |
No | |
7.4 to 8.1 | |
Bovine | |
-20°C |
Liquid | |
Sterile | |
12.4mg/dL | |
296mOsm | |
7.2 to 8.5g/dL | |
0.2μm Filtration |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title