Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Bovine Albumin, Path 4

Used as medium in density gradient centrifugation or as an addition to tissue culture growth medium

$386.50

Specifications

Format Solution
For Use With (Application) Immunoassays, Mammalian Cell Culture
Name Bovine Albumin, Path 4
pH Range 7
Source Bovine
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
ICN810101
View Documents Promotion Details
MP Biomedicals™
08810101
50mL
Each for $386.50
Add to cart
 
Description

Description

  • 35% Albumin solution
  • Sterile

Immunoassays; Mammalian Cell Culture

Specifications

Specifications

Solution
Bovine Albumin, Path 4
Bovine
Immunoassays, Mammalian Cell Culture
7
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit