Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Casein Hydrolysate

5% Sterile Solution

$54.10 - $87.20

Specifications

Format Liquid
Name Casein Hydrolysate Enzymatic
Storage Requirements 15° to 30°C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
ICN10486490
View Documents
MP Biomedicals™
0210486490
500mL
Each for $87.20
Add to cart
 
ICN10486480
View Documents
MP Biomedicals™
0210486480
100mL
Each for $54.10
Add to cart
 
Description

Description

  • Format: Liquid
  • Vitamin Free
  • Sterile
  • Used as nutritional source in microbial media

Specifications

Specifications

Liquid
Casein Hydrolysate Enzymatic
15° to 30°C
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit