Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Cholesterol Oxidase from Streptomyces species

Cholesterol oxidase (CHOD) catalyzes the first step in cholesterol catabolism

$63.50 - $413.00

Specifications

Color Dark Yellow
Concentration ≈15U/mg
Product Type Cholesterol Oxidase
Format Lyophilized Powder
For Use With (Application) Catalyzes the first step in Cholesterol catabolism
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
ICN15067425
View Documents
MP Biomedicals™
0215067425
25U
Each for $63.50
Add to cart
 
ICN15067480
View Documents
MP Biomedicals™
0215067480
100U
Each for $211.50
Add to cart
 
ICN15067483
View Documents
MP Biomedicals™
0215067483
250U
Each for $413.00
Add to cart
 
Description

Description

CAS # : 9028-76-6

Form: Lyophilized powder

Activity: Approximately 15 units/mg solid

Stability: Stable at -20°C. A decrease in activity of approximately 10% may occur at 2 to 8°C within 6 months.

Stabilizers: Bovine serum albumin, sugars.

Isoelectric point: 5.1 ± 0.1 and 5.4 ± 0.1

Michaelis Constant: 4.3 x 10-5 M (Cholesterol)

Inhibitors: Ionic detergents, Hg++, Ag+

Optimum pH: 6.5 to 7.0

Optimum Temperature: 45° to 50°C

pH Stability: 5.0 to 10.0 (25°C, 20 hr)

Thermal Stability: below 45°C (pH 7.0, 15 minutes)

This enzyme is useful for the determination of cholesterol in serum by coupling with cholesterol esterase

Specifications

Specifications

Dark Yellow
Cholesterol Oxidase
Catalyzes the first step in Cholesterol catabolism
≈34000
Steptomyces species
≈15U/mg
Lyophilized Powder
Ionic detergents, Hg++, Ag+
6.5 to 7
0°C
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit