Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MP Biomedicals™ Collagen Type I from Calf Skin
Suitable for cell adhesion in tissue culture as well as gel formation, platelet aggregation and as a substrate for collagenase
Supplier: MP Biomedicals™ 0215002683
Description
- CAS Number: 9007-34-5
- EC Number: 232-697-4
- MDL Number: MFCD00130825
- Format: Powder
- Soluble in 0.075M sodium citrate pH 3.7 and in dilute acetic acid
For attachment of primary cultures of epithelioid cells and many other cell types
Specifications
Tumor Marker | |
250mg | |
Chorionic Gonadotropin from Human Pregnancy Urine | |
Lyophilized |
3.7 | |
Calf Skin | |
-20°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction