Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Collagen Type I from Calf Skin

Suitable for cell adhesion in tissue culture as well as gel formation, platelet aggregation and as a substrate for collagenase

$45.45 - $387.00

Specifications

Format Lyophilized
For Use With (Application) Tumor Marker
pH 3.7
Source Calf Skin
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
ICN15002610
View Documents
MP Biomedicals™
0215002610
10mg
Each for $45.45
Add to cart
 
ICN15002680
View Documents
MP Biomedicals™
0215002680
100mg
Each for $227.50
Add to cart
 
MP215002683
View Documents
MP Biomedicals™
0215002683
250mg
Each for $387.00
Add to cart
 
Description

Description

  • CAS Number: 9007-34-5
  • EC Number: 232-697-4
  • MDL Number: MFCD00130825
  • Format: Powder
  • Soluble in 0.075M sodium citrate pH 3.7 and in dilute acetic acid

For attachment of primary cultures of epithelioid cells and many other cell types

Specifications

Specifications

Lyophilized
Tumor Marker
3.7
Calf Skin
Safety and Handling

Safety and Handling

SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit