Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MP Biomedicals™ Collagenase Type III from Clostridium histolyticum
Collagenases degrades native helical collagen fibrils
Supplier: MP Biomedicals™ 0215070401
Description
- CAS Number: 9001-12-1
- EC Number: 232-582-9
- MDL Number: MFCD00130830
- Format: brown lyophilized powder
- Specific Activity: >100U/mg
- Optimum pH: 7 to 9
Collagenase has found widespread application in isolation of specific cell types from attendant connective tissue
Specifications
1 mg/mL | |
7 to 9 | |
1mg/mL of Enzyme in 0.05M TES with 0.36mM Calcium Chloride, pH 7.5 | |
Clostridium histolyticum | |
Collagenase Type III | |
Metal chelating agents such as Cysteine, EDTA or o-Phenanthroline |
Brown | |
Cell dissociation and cell lysis | |
4°C | |
1 g | |
105000 and 57400 | |
Lyophilized Solid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction