Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Collagenase Type III from Clostridium histolyticum

Collagenases degrades native helical collagen fibrils

$542.00 - $1629.00

Specifications

Color Brown
Concentration 1mg/mL
Product Type Collagenase Type III
Format Lyophilized Solid
For Use With (Application) Cell dissociation and cell lysis
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
ICN15070401
View Documents
MP Biomedicals™
0215070401
1g
Each for $542.00
Add to cart
 
ICN15070405
View Documents
MP Biomedicals™
0215070405
5 x 1g
Each for $1,629.00
Add to cart
 
Description

Description

  • CAS Number: 9001-12-1
  • EC Number: 232-582-9
  • MDL Number: MFCD00130830
  • Format: brown lyophilized powder
  • Specific Activity: >100U/mg
  • Optimum pH: 7 to 9

Collagenase has found widespread application in isolation of specific cell types from attendant connective tissue

Specifications

Specifications

Brown
Collagenase Type III
Cell dissociation and cell lysis
105000 and 57400
Clostridium histolyticum
4°C
1mg/mL
Lyophilized Solid
Metal chelating agents such as Cysteine, EDTA or o-Phenanthroline
7 to 9
1mg/mL of Enzyme in 0.05M TES with 0.36mM Calcium Chloride, pH 7.5
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit