Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Diaphorase from Porcine Heart

Diaphorase is used in a study to assess the protein-protein interactions in assembly of lipoic acid on 2-oxoacid dehydrogenases of aerobic metabolism

$47.50 - $200.50

Specifications

Color Pale Yellow
Product Type Diaphorase
Format Lyophilized Powder
For Use With (Application) Study the assess the Protein-Protein interactions in assembly of Lipoic Acid on the 2-oxoacid Dehydrogenases of aerobic metabolism
Source Procine heart
View More Specs

Products 2
Catalog Number Mfr. No. Concentration Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Concentration Quantity Price Quantity & Availability  
ICN15084405
View Documents
MP Biomedicals™
0215084405
25U/mg 5mg
Each for $200.50
Add to cart
 
ICN15084401
View Documents
MP Biomedicals™
0215084401
≈40U/mg 1mg
Each for $47.50
Add to cart
 
Description

Description

  • CAS Number: 9001-18-7
  • EC Number: 232-587-6
  • MDL Number: MFCD00130947
  • Physical Appearance: Pale yellow, freeze dried powder
  • Specific Activity: Approx. 10U/mg protein
  • Format: Freeze-dried powder in presence of sucrose
  • Soluble in distilled water or dilute buffer

Specifications

Specifications

Pale Yellow
Lyophilized Powder
Procine heart
Diaphorase
Study the assess the Protein-Protein interactions in assembly of Lipoic Acid on the 2-oxoacid Dehydrogenases of aerobic metabolism
0°C
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit