MP Biomedicals FastPrep -24™ Instrument
High-speed, digitally controlled benchtop homogenizer for the lysis of biological samples
Uses a unique, optimized motion to disrupt cells through multidirectional, simultaneous beating of specialized lysing matrix beads on the sample material.
$12780.43
  • Products 1
  • Description & Specifications
Products
Catalog Number Mfr. No. Description Price Quantity Availability  
MP116004500
MP Biomedicals
116004500
Benchtop
Each for $12,780.43
Check Availability
Add to Cart
Descriptions & Specifications
Related Products