Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Fetal Bovine Serum, CellECT™ Silver

Aseptically collected via cardiac puncture

Specifications

Certifications/Compliance USDA
Color Golden Brown
Concentration 3.1 to 4.4g/dL
Format Liquid
Product Type Animal Serum Culture Media
View More Specs

Products 1
Catalog Number Mfr. No. Sterilization Method Price Quantity & Availability  
Catalog Number Mfr. No. Sterilization Method Price Quantity & Availability  
ICN2916154
View Documents
MP Biomedicals™
092916154/DEL
Filtration This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

  • Collected from a USDA regulated and monitored abbatoir by technicians who work onsite
  • Careful handling minimizes cell lysis and other detrimental factors, thereby reducing possible lysozyme and hemoglobin contamination
  • Supplied sterile, with no detectable mycoplasma or viral infection
  • Endotoxin level: slightly more than 1 ng/mL
Specifications

Specifications

USDA
3.1 to 4.4g/dL
Animal Serum Culture Media
Used to supplement cell culture media for the optimal growth of many cell types, effective in promoting and sustaining growth of vertebrate mammalian and insect cells
No
6.7 to 7.8
Sterile
USDA Inspected
Golden Brown
Liquid
Slightly exceed 1ng/mL
27mg/dL
280 to 325mOsm/kg
1 x 500mL
-20°C
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit