Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Fixed Control Slide for Mycoplasma Hoechst Stain Kit

For quality control use in qualitative detection of mycoplasma contamination in cell cultures by DNA fluorochrome staining

Manufacturer:  MP Biomedicals™ 093030400

Catalog No. MP093030400


Add to cart

Description

Description

Recommended Storage

Store at room temperature

Specifications

Specifications

Store at room temperature
1 Pk.
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit