Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Freund's Adjuvant, Complete

Enhances the antibody response of animals injected with antigen. Made by a modification of Freund's technique

$173.60

Specifications

Color Yellow and Hazy
Format Liquid
Formulation Paraffin oil 80 to 90%, Arlacel A 10 to 20%, heat-killed Mycobacterium butyricum 0.1 to 1.5%
For Use With (Application) Used to enhance the antibody response of animals injected with antigen
Product Name Freund's complete adjuvant
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
ICN642851
View Documents
MP Biomedicals™
08642851
50mL Each for $173.60
Add to cart
 
Specifications

Specifications

Yellow and Hazy
Paraffin oil 80 to 90%, Arlacel A 10 to 20%, heat-killed Mycobacterium butyricum 0.1 to 1.5%
Freund's complete adjuvant
Liquid
Used to enhance the antibody response of animals injected with antigen
2° to 8°C
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit