Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MP Biomedicals Glucose-6-phosphate Dehydrogenase from Leuconostoc mesenteriodes
Catalyzes the conversion of glucose-6-phosphate to 6-phosphogluconolacetone
Manufacturer: MP Biomedicals 0215118601
Description
Cytosolic enzyme in the pentose phosphate pathway, a metabolic pathway that supplies reducing energy to cells (such as erythrocytes) by maintaining the level of the co-enzyme nicotinamide adenine dinucleotide phosphate (NADPH).
- Format: White milky liquid
- Molecular Weight: 104,000 (two subunits of ~55,000) (Lit.)
- Specific Activity: ≥250units/mg protein
Specifications
Glucose-6-phosphate Dehydrogenase | |
≥250U/mg | |
Acyl-CoA, Metal ions | |
7.8 | |
1kU | |
4°C |
Liquid | |
White | |
Lueconostoc mesenteroides | |
104000 | |
Enzyme in Pentose Phosphate pathway |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title