Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Glucose-6-phosphate Dehydrogenase from Leuconostoc mesenteriodes

Catalyzes the conversion of glucose-6-phosphate to 6-phosphogluconolacetone

$209.50 - $675.50

Specifications

Color White
Concentration ≥250U/mg
Product Type Glucose-6-phosphate Dehydrogenase
Format Liquid
For Use With (Application) Enzyme in Pentose Phosphate pathway
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
ICN15118601
View Documents
MP Biomedicals™
0215118601
1kU
Each for $209.50
Add to cart
 
ICN15118605
View Documents
MP Biomedicals™
0215118605
5kU
Each for $675.50
Add to cart
 
Description

Description

Cytosolic enzyme in the pentose phosphate pathway, a metabolic pathway that supplies reducing energy to cells (such as erythrocytes) by maintaining the level of the co-enzyme nicotinamide adenine dinucleotide phosphate (NADPH).

  • Format: White milky liquid
  • Molecular Weight: 104,000 (two subunits of ~55,000) (Lit.)
  • Specific Activity: ≥250units/mg protein

Specifications

Specifications

White
Glucose-6-phosphate Dehydrogenase
Enzyme in Pentose Phosphate pathway
104000
Lueconostoc mesenteroides
≥250U/mg
Liquid
Acyl-CoA, Metal ions
7.8
4°C
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit