Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Goat Serum: Normal Pool, Lyophilized, Sterile Filtered

Catalog No. ICN642921
Click to view available options
Quantity:
50 mL

Sterile, filtered, Lyophilized Serum

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2° to 8°C
Species Goat
Product Type Goat Serum
Quantity 50 mL
Sterility Sterile
Sterilization Method Filtration
For Use With (Application) Immunocytochemistry and blocking
Lyophilized Yes
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.