Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Histone from Calf Thymus

Arginine-rich subgroup

$82.00 - $163.00

Specifications

Format Powder
Name Histone
Source Calf
Storage Requirements 2° to 8°C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
ICN10072610
View Documents Promotion Details
MP Biomedicals™
0210072610
10mg
Each for $82.00
Add to cart
 
ICN10072625
View Documents Promotion Details
MP Biomedicals™
0210072625
25mg
Each for $163.00
Add to cart
 
Description

Description

Specifications

Specifications

Powder
Histone
Calf
2° to 8°C
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit