Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Human Growth Factors and Supplements

Suitable for supplementing cell cultures and investigating various metabolic processes

Specifications

Cross Reactivity Human
For Use With (Application) Supplementing cell cultures and investigating various metabolic processes
Lyophilized Yes
Storage Requirements -20°C
View More Specs

Products 1
Catalog Number Mfr. No. Source Type Format Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Source Type Format Quantity Price Quantity & Availability  
ICN15350880
View Documents
MP Biomedicals™
15350880
Recombinant, expressed in E. coli. Epidermal Growth Factor Recombinant 100μg This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

Specifications

Specifications

Human
Supplementing cell cultures and investigating various metabolic processes
Yes
-20°C
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit