Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Human Serum, Type AB

Golden Yellow Liquid. Proven to grow many human cell lines at a faster rate and with a smaller percentage of serum than mixed blood group serum.

$347.00

Specifications

Format Liquid
Sterility Sterile
Source Human
Storage -20°C
View More Specs

Products 1
Catalog Number Mfr. No. Hemoglobin Concentration pH Protein Content Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Hemoglobin Concentration pH Protein Content Quantity Price Quantity & Availability  
ICN2938249
View Documents
MP Biomedicals™
092938249
≤26mg/dL 7 to 8 5.5 to 7.5g/dL 100mL Each for $347.00
Add to cart
 
Specifications

Specifications

Liquid
Sterile
Human
-20°C
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit