Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Human Serum, Type AB, Normal Pool

Used as a growth supplement for cell culture media and in other research applications.

$314.50 - $332.50

Specifications

Certifications/Compliance U.S. regulations
Format Liquid
Sterility Sterile
For Use With (Application) Grow many Human cell lines at a faster rate
Hemoglobin Concentration ≤26mg/dL
View More Specs

Products 2
Catalog Number Mfr. No. Color Product Type Protein Content Price Quantity & Availability  
Catalog Number Mfr. No. Color Product Type Protein Content Price Quantity & Availability  
ICN2930949
View Documents
MP Biomedicals™
092930949
Straw Yellow to Golden Brown Human Type AB Serum, (HBsAG), HIV-free 5 to 8.3g/dL Each for $332.50
Add to cart
 
ICN2931949
View Documents
MP Biomedicals™
092931949
Golden Brown Human Type AB Serum, HCV-free 5.5 to 7.5g/dL Each for $314.50
Add to cart
 
Specifications

Specifications

U.S. regulations
Sterile
≤26mg/dL
263 to 323mOsm/kg
100mL
0.1μm Filtration
Liquid
Grow many Human cell lines at a faster rate
No
7 to 8
Human
-20°C
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit