Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Hyaluronidase, Ovine testes

Source: Ovine Testes

Specifications

Color White
Format Lyophilized
For Use With (Application) Drug dispersion and delivery
Inhibitors Fe2+ and Fe3+ are inhibitory as are Mn2+ and Cu2+
Molecular Weight (g/mol) ≈55
View More Specs

Products 1
Catalog Number Mfr. No. Concentration Product Type Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Concentration Product Type Quantity Price Quantity & Availability  
ICN15127480
View Documents
MP Biomedicals™
0215127480
>1500 U/mg Hyaluronidase 100 mg This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

Hyaluronidase catalyzes the random hydrolysis of 1,4-linkages between 2-acetamido-
2-deoxy- b-D-glucose and D-glucose residues in hyaluronate.

  • CAS # : 37326-33-3
  • Synonyms: Hyaluronate 4-glycanohydrolase; hyaluronate lyase; hyaluronoglucosaminidase; E.C. 3.2.1.35
  • Physical Description: Off white crystalline powder
  • Unit Definition: That amount of enzyme causing the same turbidity reduction as the International Unit (I.U.) as compared with the international standard.
  • Molecular weight: Borders and Raftery (1968) reported a molecular weight of 61,000. Khorlin, et. al.(1973) report four subunits of 14,000 each and a total molecular weight of 55,000.
  • Solubility: Dissolves readily at 5mg/mL in 0.02M sodium phosphate/0.45% sodium chloride/NaOH/0.01% BSA pH 6.9 to give a clear colorless solution .
  • Optimum pH: 4.5 - 6 (DeSalequi et al. 1967).
  • Extinction coefficient: = approximately 8.
  • Inhibitors: Fe2+ and Fe3+ are inhibitory as are Mn2+ and Cu2+(Warren et al. 1962).
  • Specificity: Testicular hyaluronidase hydrolyzes the endo-N-acetylhexosaminic bonds of hyaluronic acid and chondroitin sulfuric acids A and C (but not B), primarily to tetrasaccharide residues (Ludowieg et al. 1961). Monosaccharides are not liberated (Rappaport et al. 1951).
  • Stabilizers: Yang and Srivastav (1975) report that sodium chloride acts as a stabilizer .

Specifications

Specifications

White
Drug dispersion and delivery
≈55
Ovine testes
Lyophilized
Fe2+ and Fe3+ are inhibitory as are Mn2+ and Cu2+
4.5 to 6
Safety and Handling

Safety and Handling

SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit