Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MP Biomedicals™ Intrinsic Factor
Derived from Porcine Pyloric Mucous Lining
Supplier: MP Biomedicals™ 0210205650
Description
- Format: Tan powder
- Specific Activity: Standard quality of contains at least 50 Coli units per mg (total binding)
Absorption of vitamin B12
Specifications
Vitamin B12 Absorption | |
50mg | |
Powder |
Intrinsic Factor from Porcine Pyloric Mucous Lining | |
0°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction