Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Intrinsic Factor

Derived from Porcine Pyloric Mucous Lining

$199.00 - $2554.00

Specifications

For Use With (Application) Vitamin B12 Absorption
Name Intrinsic Factor from Porcine Pyloric Mucous Lining
Storage Requirements 0°C
Form Powder
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
ICN10205650
View Documents
MP Biomedicals™
0210205650
50mg
Each for $199.00
Add to Cart
 
ICN10205680
View Documents
MP Biomedicals™
0210205680
100mg
Each for $404.00
Add to Cart
 
ICN10205690
View Documents
MP Biomedicals™
0210205690
500mg
Each for $1,275.00
Add to Cart
 
ICN10205691
View Documents
MP Biomedicals™
0210205691
1g
Each for $2,554.00
Add to Cart
 
Description

Description

  • Format: Tan powder
  • Specific Activity: Standard quality of contains at least 50 Coli units per mg (total binding)

Absorption of vitamin B12

Specifications

Specifications

Vitamin B12 Absorption
Intrinsic Factor from Porcine Pyloric Mucous Lining
0°C
Powder
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit