Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MP Biomedicals Lactalbumin
Nutritional supplement possessing high protein content and amino acid balance
Manufacturer: MP Biomedicals 0210212801
Description
- Format: Off-white powder
- Albumin fraction of milk rich in lysine
Specifications
Protein (N x 6.38), as is basis 80%, Lactose 1%, Mineral Salts 3%, Lipid Matter 7%, Moisture 5%, Heavy Metals 10 ppm, Sodium 0.1%, pH (10% suspension) 4.5 to 5.5 | |
Lactalbumin | |
15°C to 30°C |
Powder | |
450 g |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title