Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Lactalbumin

Nutritional supplement possessing high protein content and amino acid balance

Manufacturer:  MP Biomedicals™ 0210212805

 View more versions of this product

Catalog No. ICN10212805

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

  • Format: Off-white powder
  • Albumin fraction of milk rich in lysine

Specifications

Specifications

2260 g
Lactalbumin
Powder
15°C to 30°C
Protein (N x 6.38), as is basis 80%, Lactose 1%, Mineral Salts 3%, Lipid Matter 7%, Moisture 5%, Heavy Metals 10 ppm, Sodium 0.1%, pH (10% suspension) 4.5 to 5.5
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit