Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Linbro™ Tissue Culture Plates

Specifications

Color Clear
Sterility Sterile
For Use With (Application) Ideal for a variety of applications including cell cloning and metabolic labeling procedures
Closure Included With Lid
Material Polystyrene
View More Specs

Products 1
Catalog Number Mfr. No. Culture Area Capacity (Metric) Dimensions (L x W x H) Price Quantity & Availability  
Catalog Number Mfr. No. Culture Area Capacity (Metric) Dimensions (L x W x H) Price Quantity & Availability  
ICN7605305
View Documents
MP Biomedicals™
097605305
4.5 cm2 7.5 mL 111 x 84 x 22 mm This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

Specifications

Specifications

Clear
Ideal for a variety of applications including cell cloning and metabolic labeling procedures
Polystyrene
Sterile
With Lid
Tissue Culture Plate
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit