Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Lysing Matrix G

Tubes for optimal lysis used with FastPrep™ instruments

$121.50 - $241.50

Specifications

For Use With (Application) Nucleic Acid isolation and purification, molecular biology, gene expression, microbiological analysis
Matrix Lysing matrix G
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MP116916050
View Documents
MP Biomedicals™
116916050
50 x 2 mL Tubes
Each for $121.50
Add to cart
 
MP116916100
View Documents
MP Biomedicals™
116916100
100 x 2 mL Tubes
Each for $241.50
Add to cart
 
Description

Description

Silicon carbide has relatively low density and high strength and hardness, and therefore is effective at breaking tough, hard, or brittle cell membranes. Useful in applications such as isolation of intact organelles, and super-molecular structures where lysis conditions necessitate high impaction but low shearing.

  • Impact-resistant 2mL tube contains 1.6mm Silicon Carbide particles and 2mm glass beads
  • With brown caps
  • Specific Gravity: Silicon Carbide: 3.20; Glass beads: 2.55

Nucleic Acid Isolation and Purification

Specifications

Specifications

Nucleic Acid isolation and purification, molecular biology, gene expression, microbiological analysis
Lysing matrix G
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit