Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Lysing Matrix Y, 96-Well Rack
GSA_VA

Optimized for the FastPrep™-96 Instrument and are included in the FastDNA™-96 Soil Microbe DNA Kit, the FastDNA™-96 Fungal/Bacterial DNA Kit, and the FastDNA™-96 Fecal DNA Kit

$202.00 - $1338.00

Specifications

For Use With (Application) cracking of very tough cell walls
For Use With (Equipment) FastPrep-96
View More Specs

Products 2
Catalog Number Mfr. No. Includes Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Includes Quantity Price Quantity & Availability  
MP116960001
View Documents
MP Biomedicals™
116960001
1.2mL cluster-strip tube pre-filled w/0.5mm dia. Yttria-Stabilized Zirconium Oxide beads 96 x 1.2 mL
Each for $202.00
Add to cart
 
MP116960010
GSA_VA
View Documents
MP Biomedicals™
116960010
1.2mL cluster-strip tube pre-filled w/0.5mm dia. Yttria-Stabilized Zirconium Oxide beads 10 x 2 mL
Each for $1,338.00
Add to cart
 
Description

Description

  • Each 96 deep well rack comes with 1.2mL cluster-strip tube and cap assemblies.
  • Each tube is pre-filled with 0.5mm diameter Yttria-Stabilized Zirconium Oxide beads.
  • Y2O3 + ZrO2 has high density and has extremely high hardness making this matrix ideal for cracking of very tough cell walls, while it's smooth surface minimizes shearing.
  • These Lysing Matrix racks are optimized for the FastPrep™-96 Instrument and are included in the FastDNA™-96 Soil Microbe DNA Kit, the FastDNA™-96 Fungal/Bacterial DNA Kit, and the FastDNA™-96 Fecal DNA Kit
Specifications

Specifications

cracking of very tough cell walls
FastPrep-96
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit