Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MP Biomedicals™ Malate Dehydrogenase, From Porcine Heart (Mitochondrial)
Used for enzymatic determination of L-malate and gluamate oxaloacetate transaminase (GOT)
Supplier: MP Biomedicals™ 0216004950
Description
Malate Dehydrogenase, [9001-64-3]
- CAS Number: 9001-64-3
- EC Number: 232-622-5
- Specific Activity: ∽1200u/mg protein
- Format: Powder
Specifications
Lyophilized Powder | |
White | |
0°C | |
50 kU |
≈1200 U/mg | |
Enzymatic determination of L-malate and Gluamate Oxaloacetate Transaminase | |
Porcine heart | |
Malate Dehydrogenase |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title