Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MP Biomedicals Merrifield's Peptide Resin
Sieve Size of 200 to 400 mesh or 40 to 80µm
Manufacturer: MP Biomedicals 0215160825
Specifications
25g | |
White Powder | |
Merrifields Peptide Resin |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title