Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Merrifield's Peptide Resin

Sieve Size of 200 to 400 mesh or 40 to 80μm

$721.00

Specifications

Format Powder
Product Type Merrifields Peptide Resin
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
ICN15160880
View Documents
MP Biomedicals™
0215160880
100 g
Each for $721.00
Add to cart
 
Description

Description

Specifications

Specifications

Powder
Merrifields Peptide Resin
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit