Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Other Growth Factors and Supplements

Suitable for supplementing cell cultures and investigating various metabolic processes

Specifications

For Use With (Application) Supplementing cell cultures and investigating various metabolic processes
View More Specs

Products 4
Catalog Number Mfr. No. Source Product Type Quantity Content And Storage Price Quantity & Availability  
Catalog Number Mfr. No. Source Product Type Quantity Content And Storage Price Quantity & Availability  
ICN15233025
View Documents
MP Biomedicals™
0215233025
Bovine Brain Growth Supplement 25 mg 2° to 8°C Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area. N/A
ICN15984010
View Documents
MP Biomedicals™
0215984010
Naja naja kaouthia (Thailand Cobra) Venom Growth Factor 10 μ g 0° to 5°C Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area. N/A
ICN16003701
View Documents
MP Biomedicals™
16003701
Bovine Pituitary Gland Growth Factor 10 μ g -20°C Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area. N/A
ICN19113080
View Documents
MP Biomedicals™
19113080
Growth Factor Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area. N/A
Description

Description

Specifications

Specifications

Supplementing cell cultures and investigating various metabolic processes
Safety and Handling

Safety and Handling

ShelfLife : 1 Month

SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit