Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MP Biomedicals™ Pectinase from Aspergillus niger
$48.75 - $222.00
Specifications
Concentration | >1000 U/g |
---|---|
Color | Beige |
For Use With (Application) | Used in plant protoplast preparation to digest cell wall prior to organelle isolation |
Content And Storage | 4°C |
Source | Aspergillus niger |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
ICN19897910
|
MP Biomedicals™
0219897910 |
10 kU |
Each for $73.50
|
|
|||||
ICN19897925
|
MP Biomedicals™
0219897925 |
25 kU |
Each for $153.00
|
|
|||||
ICN19897980
|
MP Biomedicals™
0219897980 |
100 kU |
Each for $222.00
|
|
|||||
ICN19897901
|
MP Biomedicals™
0219897901 |
1 kU |
Each for $48.75
|
|
|||||
ICN19897905
|
MP Biomedicals™
0219897905 |
5 kU | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
Description
Pectinase is a macerating enzyme comprised of pectintranseliminase, polygalacturonase, pectinesterase, xylanase, hemicellulases and cellulases.
- Lyophilized
- Activity: >1000 u/gm
Specifications
>1000 U/g | |
Used in plant protoplast preparation to digest cell wall prior to organelle isolation | |
Aspergillus niger | |
Lyophilized Powder |
Beige | |
4°C | |
Pectinase |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title