Learn More
MP Biomedicals Pepsin From Porcine Stomach Mucosa
Pepsin cleaves peptides with an aromatic acid on either side of the peptide bond
$113.40 - $576.50
Specifications
Format | Powder |
---|---|
For Use With (Application) | Molecular Biology, Proteomics |
Name | Pepsin |
pH Range | 2.5 to 4.5 |
Source | Porcine |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
ICN10259850
|
MP Biomedicals
0210259850 |
50g |
Each for $113.40
|
|
ICN10259890
|
MP Biomedicals
0210259890 |
500g |
Each for $576.50
|
|
Description
Pepsin contains the “cathepsin” component which has milk curdling activity. It has a broad range of substrate activity and demonstrates an esterase acitivity.
- CAS Number: 9001-75-6
- EC Number: 232-629-3
- MDL Number: MFCD00081840
- Format: off white lyophilized powder
- Specific activity: ≥1:10000
Used in preparation of Fab fragments from antibodies and to hydrolyze dry cervical samples in mice
Specifications
Powder | |
Pepsin | |
Porcine |
Molecular Biology, Proteomics | |
2.5 to 4.5 | |
15° to 30°C |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.