Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Peroxidase, From Horseradish

From Horseradish

$600.00

Specifications

For Use With (Application) ELISA, immunoblotting, immunohistochemistry
Inhibitors Cyanide, Sulfide, p-Aminobenzoic Acid, Azide, Cyclopropanone, L-Cystine, Dichromate, Ethylenethiourea, Hydroxylamine, Sulfite, Vanadate and the Divalent Metals Cd, Co, Cu, Fe, Mn, Ni, Pb
Source Horseradish
Format Lyophilized
Product Type Peroxidase
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
ICN15003810
View Documents
MP Biomedicals™
0215003810
10kU
Each for $600.00
Add to cart
 
Description

Description

Peroxidase, [9003-99-0]

  • Lactoperoxidase
  • A salt-free, freeze dried powder
  • Activity: greater than 100 purpurogallin units/mg solid.
  • Unit Definition: That amount of enzyme causing the production of one milligram of purpurogallin in 20 seconds at 20°C.

Specifications

Specifications

ELISA, immunoblotting, immunohistochemistry
Horseradish
Peroxidase
Cyanide, Sulfide, p-Aminobenzoic Acid, Azide, Cyclopropanone, L-Cystine, Dichromate, Ethylenethiourea, Hydroxylamine, Sulfite, Vanadate and the Divalent Metals Cd, Co, Cu, Fe, Mn, Ni, Pb
Lyophilized
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit