Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Plant Culture Nutrient Mixtures

$57.00

Specifications

Storage Requirements 0° to 5°C
Quantity 100mL
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
ICN2625149
View Documents
MP Biomedicals™
092625149
100mL
Each for $57.00
Add to cart
 
Description

Description

Each formulation is prepared according to the original specification and is of single strength, except where noted.

1Store at 0° to 5°C.
2Protect from light.

Specifications

Specifications

0° to 5°C
100mL
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit