Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Poly-L-Lysine Hydrobromide Attachment Factors: 30,000-70,000 MW

Catalog No. p-4906918
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Sterility:
Non-sterile
Sterile

Facilitate attachment, adherence, and spreading in the culture of adherent cells

Available nonsterile or gamma-irradiated.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage -20°C
Product Type Poly-L-Lysine
Sterility Non-sterile
Quantity 10 mg
Source Synthetic
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.