Learn More
MP Biomedicals™ Rabbit Serum
Sterile, Frozen Brown Solution. Combine with BSA to use as a blocking serum in immunocytochemistry and immunohistochemistry.
Supplier:  MP Biomedicals™ 0219135720
 View more versions of this product
Catalog No. ICN19135720
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Description
Description
Specifications
Specifications
-20°C
Liquid
20 mL
Sterile
Blocking agent in immunohistochemistry culture media component
Rabbit
Rabbit Serum
4 to 7 g/dL
Membrane-filtered
6.5 to 8
Product Suggestions
Product Suggestions