Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Raffinose Powder

Catalog No. p-4474965
Click to view available options
Quantity:
227 g
454 g

Pentahydrate powder is 98% pure

  • White powder

Yeast cell culture, Gene expression

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 15°C to 30°C
Additive D-(+)-Raffinose Pentahydrate
Form Powder
For Use With (Application) Yeast cell culture, gene expression
Quantity 454 g
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.