Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ RNA PLUS

Used for RNA isolation from tissues and cells of human, animal, plant, yeast and bacterial origin

Manufacturer:  MP Biomedicals™ 11RNAPLUS2

Catalog No. 11-RNA-PLUS2


Add to cart

Description

Description

The RNA is suitable for all downstream applications including RT-PCR, hybridization, Northerns, polyA + mRNA isolation, in vitro transcription. RNA-PLUS™ represents an improvement on Chomczynski's procedure for single step RNA isolation providing higher efficiency removal of DNA and proteins. The method promotes formation of complexes of RNA with guanidinium and water molecules and abolishes hydrophilic interactions of DNA and proteins during sample extraction. RNAs obtained with RNA-PLUS™ are not degraded and are free of proteins and DNA. 1mL of reagent is sufficient for 50mg tissue or 5 x 106 cells.

Specifications

Specifications

Nucleic Acid Isolation & Purification; Sample Lysis & Preparation
RNAPLUS
100mL
Store at 2-8°C.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit