Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Streptomycin Sulfate

Serve variety of purposes in cell culture, including selection, elimination of mycoplasma and microbial contamination, and prophylaxis

$41.80 - $105.40

Specifications

Format Powder
Solution Type Streptomycin Sulfate
Storage 4°C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
ICN19454125
View Documents
MP Biomedicals™
0219454125
25g
Each for $41.80
Add to cart
 
ICN19454180
View Documents
MP Biomedicals™
0219454180
100g
Each for $105.40
Add to cart
 
Description

Description

All antibiotics are intended for research use only. They are not for therapeutic use in humans or animals.

Antibiotic effective against most gram negative and gram positive bacteria.

Specifications

Specifications

Powder
Streptomycin Sulfate
4°C
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit