Learn More
MP Biomedicals Titertek Finnpipette Multistepper Pipet Tips
Hydrophobic tips with very low coefficient of friction
Manufacturer: MP Biomedicals 0977943C2
Description
Specifically for the Titertek/Labsystems 8-channel Multistepper pipette
- Made from virgin polypropylene material
- Comes with robot-compatible (large-micron) hydrophobic filters to eliminate instrument contamination
- 96 tips per rack
- Volume: 1500µL
Specifications
1500 μL | |
Natural | |
Bulk |
Non-sterile | |
PP (Polypropylene) |