Learn More
MP Biomedicals™ Transport Factors
Supplier:  MP Biomedicals™ 08823411
 View more versions of this product
Catalog No. ICN823411
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Transport factors are proteins which enable small metabolites to be carried in usable form to cells. Transferrin binds iron and makes it available to cells in a recognizable form.
†Contains <1.0% iron.
Specifications
Specifications
-20°C
7.24
Pooled Human Plasma
Transferrin
1 g