Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Transport Factors

$1131.00

Specifications

Content And Storage -20°C
Product Type Transferrin
Quantity 1 g
Source Pooled Human Plasma
View More Specs

Products 2
Catalog Number Mfr. No. pH Price Quantity & Availability  
Catalog Number Mfr. No. pH Price Quantity & Availability  
ICN823431
View Documents
MP Biomedicals™
08823431
6.8
Each for $1,131.00
Add to cart
 
ICN823411
View Documents
MP Biomedicals™
08823411
7.24 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

Transport factors are proteins which enable small metabolites to be carried in usable form to cells. Transferrin binds iron and makes it available to cells in a recognizable form.

Specifications

Specifications

-20°C
Transferrin
1 g
Pooled Human Plasma
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit