Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MP Biomedicals™ Voltage Converter for SuperFastPrep-1™
For use with SuperFastPrep-1™
Supplier: MP Biomedicals™ 116011520
Description
- 100W Transformer
- 220 to 240VAC to 100 to 120VAC
- For SuperFastPrep™
Specifications
Voltage Converter | |
SuperFastPrep-1™ | |
1 Ea. | |
100W Transformer, 220-240VAC to 100-120VAC |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction