Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Water for Cell Culture, MP Biomedicals™
Click to view available options
Cell Culture Grade; 500mL
Specifications
pH | 5.0-7.0 |
CAS | 7732-18-5 |
Boiling Point | 100°C |
Molecular Formula | H2O |
Synonym | dihydrogen oxide, dihydrogen monoxide |
InChI Key | XLYOFNOQVPJJNP-UHFFFAOYSA-N |
SMILES | O |
IUPAC Name | oxidane |
Molecular Weight (g/mol) | 18.015 |
PubChem CID | 962 |
Show More |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction