Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MP Biomedicals™ Water for Cell Culture
Click to view available options
Quantity:
500mL
Specifications
Storage Requirements | Room Temperature |
Sterility | Sterile |
Molecular Weight (g/mol) | 18.015 |
Color | Undesignated |
Boiling Point | 100°C |
pH | 5.0-7.0 |
Quantity | 500mL |