Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Yeast Extract Powder

A vacuum-dried extract concentrate of Baker's yeast containing the B-complex factors of approximately three times the weight of ordinary dry yeast.

$44.90 - $282.50

Specifications

pH 5 to 5.5
Storage Requirements 15° to 30°C
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
ICN10330380
View Documents
MP Biomedicals™
0210330380
100g
Each for $44.90
Add to cart
 
ICN10330390
View Documents
MP Biomedicals™
0210330390
500g
Each for $123.30
Add to cart
 
ICN10330391
View Documents
MP Biomedicals™
0210330391
1kg
Each for $282.50
Add to cart
 
Description

Description

Specifications

Specifications

5 to 5.5
15° to 30°C
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit