Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VAI MP Hood
The MP Hood with integrated face mask is individually packaged.
$756.00
Specifications
Product Type | Hood |
---|---|
Color | White |
Disposable | Yes |
Resistance | Liquids |
Sterility | Sterile |
Catalog Number | Mfr. No. | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Price | Quantity & Availability | |||||
19-546-536
|
VAI
1700PHMS17037 |
Small/Medium |
Case of 100 for $756.00
|
|
19-546-537
|
VAI
1700PHMS17038 |
Large/X-Large |
Case of 100 for $756.00
|
|
Description
- Heavy weight, non-woven and high quality microporous (MP) coated material
- Suited for use where maximum protection to product or wearer is required
- Excellent liquid resistance, especially to chemicals
Specifications
Hood | |
Yes | |
Sterile |
White | |
Liquids | |
Individually Packaged |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title