Learn More
Abnova™ mPEG ELISA Kit
Enzyme-linked immunosorbent assay for the quantitative measurement of methoxy-PEG in serum and plasma.
Manufacturer:  Abnova™ KA4898
Catalog No. 89-032-275
Add to cart
Description
Description
  • Used for the in vitro quantitative determination of human SOD1 in human serum, human plasma, cell lysate, culture supernatants and buffered solution
  • Assay will recognize native and recombinant human IL-9
Specifications
Specifications
0.3125 to 20 ng/mL
mPEG ELISA Kit is an enzyme-linked immunosorbent assay for the quantitative measurement of methoxy-PEG in serum and plasma.
5 kDa mPEG-amine Standard
For research use only (RUO)
Store the kit at -20°C. For best results, store the components as described in the protocol. Avoid repeated freezing and thawing.
1 Kit
HRP
Quantification
mPEG ELISA Kit
100 μL
Colorimetric
Plasma, Serum